Learn More
Description
Specifications
Specifications
| Concentration | Concentration on Label |
| Formulation | In 150mM NaCl, 4mM sodium acetate buffer, pH 5.3. |
| Purification Method | Single band by SDS-PAGE |
| Quantity | 100 μg |
| Immunogen | Coagulation Factor XI |
| Storage Requirements | Following initial thaw, aliquot and freeze (−70°C). |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.