Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ COCO, Human, Recombinant Protein

Catalog No. 23355350UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-355-350UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-355-350UG Supplier MilliporeSigma™ Supplier No. 23355350UG
Only null left
Add to Cart
Add to Cart
Recombinant, human COCO consisting of amino acids 23-189 and expressed in E. coli with an N-terminal methionine. Inhibits BMP-4 induced activity in MC3T3-E1 mouse osteoblastic cells. ED 50 = 0.15-0.75μg/mL. Human COCO, at the amino acid sequence level, is 60% and 24% identical with mouse and Xenopus COCO, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.2μM sterile-filtered Acetonitrile, TFA, BSA.
Preparation Method Reconstitute to 200μg/mL in sterile 4mM HCI containing at least 0.1% human or bovine serum albumin.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen COCO
Storage Requirements Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.