Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C1-Esterase Inhibitor, Human, Protein

Catalog No. 2048831MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-488-31MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-488-31MG Supplier MilliporeSigma™ Supplier No. 2048831MG
Add to Cart
Add to Cart
Native, human C1-esterase. Single-chain glycoprotein present in normal human serum at 180μg/mL. Involved in regulation of complement, coagulation, fibrinolytic and contact (kinin-forming) systems of circulating blood plasma. The only known inhibitor of activated C1r and C1s complement enzymes. The presence of either hereditary or acquired deficiencies of C1 inhibitor produce susceptibility to recurrent attacks of angioedema.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, pH 7.4, sterile-filtered through a 0.22μM filter.
Purification Method SDS-PAGE
Quantity 1 mg
Immunogen Complement C1-Esterase Inhibitor
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.