Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C1s, Human, Activated, Two-Chain Form, Protein

Catalog No. 204879250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-487-9250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-487-9250UG Supplier MilliporeSigma™ Supplier No. 204879250UG
Add to Cart
Add to Cart
Native, human, activated C1s complement component. Present in human serum at 31μg/mL. Nonactivated C1s is found in circulating blood plasma as a Ca2+-dependent dimer in association with one C1q molecule and two C1r molecules to form the first component of complement (C1). Ensuing C1q binding to classical complement pathway activators, each C1r protein is cleaved to form activated C1r enzyme. Dimeric C1r enzyme cleaves, and thus activates, each C1s molecule into two disulfide-linked fragments of M.W. 59kDa and 28kDa. The 28kDa peptide contains the C1s enzymatic active site. Activated C1s continues activation via classical pathway by cleaving, and thus activating, C2 and C4.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 130mM NaCl, 50mM sodium phosphate, pH 7.2.
Purification Method SDS-PAGE
Quantity 250 μg
Immunogen Complement C1s
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Structural Form Two-Chain Form
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.