Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C2, Human, Protein

Catalog No. 20488250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-488-250UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-488-250UG Supplier MilliporeSigma™ Supplier No. 20488250UG
Add to Cart
Add to Cart
Native, human C2 complement component. Single chain glycoprotein present in normal human serum at concentration of 20μg/mL. Classical pathway activation results in assembly of the C1,C4b complex on target surfaces. Subsequent to binding of the C4b component to this complex, C2 is cleaved by the activated C1s subcomponent of C1 into C2a, the 63kDa C-terminal fragment, and C2b, the 30kDa N-terminal fragment. The smaller C2b fragment is released while the larger C2a fragment remains associated with the C1,C4b complex to form the C4b,C2a classical pathway C3 convertase enzyme. C2 concentration is based on functional activity measurements relative to a normal human serum standard pool.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, pH 6.0.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen Complement C2
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.