Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C3a, Human, Protein

Catalog No. 20488150UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-488-150UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-488-150UG Supplier MilliporeSigma™ Supplier No. 20488150UG
Add to Cart
Add to Cart
Native, human C3a complement component. Single chain, 77 amino acid peptide. Activation of either complement pathway can result in formation of C3-cleaving enzymes (C3 convertases) on target surfaces. The C3 convertase enzymes of either complement activation pathway will cleave the C3 α-chain at peptide bond 77 resulting in the production and release of the C3a peptide. C3a, one of the three complement-derived anaphylatoxins, expresses a wide variety of biological activities which include smooth muscle contraction, platelet and neutrophil activation and aggregation, skin wheal and flare, and immunoregulatory reactions.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, pH 7.2, sterile-filtered through a 0.22μM filter.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen Complement C3a
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.