Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C3d, Human, Protein

Catalog No. 204870100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-487-0100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-487-0100UG Supplier MilliporeSigma™ Supplier No. 204870100UG
Add to Cart
Add to Cart
Native, human C3d complement component. C3d is formed by the limited digestion of iC3b by trypsin or elastin. The C3d region of C3 contains the portion of the native C3 molecule which is capable of forming a covalent bond attachment to target surfaces under attack by complement. C3d is a polypeptide of M.W. 30kDa. The binding of C3d to CR2 (CD21) receptors present on B lymphocytes markedly enhances B cell activation initiated by a wide variety of stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, pH 7.2.
Purification Method SDS-PAGE
Quantity 100 μg
Immunogen Complement C3d
Storage Requirements Following initial thaw, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥85%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.