Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C4b, Human, Protein

Catalog No. 204897250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-489-7250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-489-7250UG Supplier MilliporeSigma™ Supplier No. 204897250UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Native, human C4b complement component. Cleavage of the C4 α-chain at peptide bond 77 by activated C1 enzyme results in the production of C4a (M.W. 8,758) and C4b (M.W. 193,000) fragments. The C4b fragment is a glycoprotein composed of the modified C4 α-chain (α') and intact β- and γ-chains. Like C3b, C4b has the transient ability to form a covalent ester bond with a variety of target cell surfaces. Once bound to the target surface, C4b becomes an essential non-enzymatic subunit of the classical pathway C3 cleaving enzyme (C4b,C2a). In addition, surface bound C4b has opsonic and immune adherence activities which are mediated via binding to the CR1 (CD35) complement receptor which is found on a variety of inflammatory cells.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, pH 7.2.
Purification Method SDS-PAGE
Quantity 250 μg
Immunogen Complement C4b
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.