Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C5, Human, Protein

Catalog No. 204888250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-488-8250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-488-8250UG Supplier MilliporeSigma™ Supplier No. 204888250UG
Only null left
Add to Cart
Add to Cart
Native, human C5 complement component. Glycoprotein composed of two non-identical subunits of M.W. 120kDa (α) and 75kDa (β) linked by disulfide bonds. Present in normal human serum at 70μg/mL. Activation of complement via either the classical or alternative pathway can result in the assembly of C5-cleaving enzymes (C5 convertases) on target surfaces. Both C5 convertases cleave the C5 α-chain at peptide bond 74 resulting in production of C5a (M.W. 11,200) and C5b fragments (M.W. 185,000). Released C5a peptide is one of the three complement-derived anaphylatoxins. C5b fragment combines with C6, C7, C8, and C9 to form lytic C5b-9 complement membrane attack complex (MAC).
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, pH 7.2.
Purification Method SDS-PAGE
Quantity 250 μg
Immunogen Complement C5
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.