Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Concanavalin A, Canavalia ensiformis, Protein

Catalog No. 234567250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-456-7250MG 250 mg
23-456-71GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-456-7250MG Supplier MilliporeSigma™ Supplier No. 234567250MG
Only null left
Add to Cart
Add to Cart
Carbohydrate-binding protein that agglutinates various animal cells. Also known to restore the growth pattern of tumor cells to that of normal cells. T lymphocyte activator. Has binding sites for metal ions, α-D-mannose, and α-D-glucose.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Salt-free.
Preparation Method To dissolve, add Con A slowly to a solution of 10mM HEPES buffered saline, 100μM CaCl2, pH 8.5. Allow the lectin to dissolve without agitation.
Quantity 250 mg
Source Canavalia ensiformis
Immunogen Concanavalin A
Storage Requirements Following reconstitution, aliquot and freeze (−20°C) or add 0.08% NaN3 and refrigerate (4°C). Stock solutions are stable for up to 6 months at −20°C or for up to 3 months at 4°C.
Regulatory Status RUO
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.