Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ CpG MethylQuest™ Protein (Recombinant GST-MBD fusion protein)
Shop All MilliporeSigma ProductsDescription
The CpG MethylQuest protein is an effective and versatile tool for the study of CpG methylation in DNA. This purified recombinant protein contains the methyl binding domain (MBD) of the mouse MBD2b protein fused to a glutathione-S-transferase protein (GST) from S japonicum . These two protein domains are separated by a linker containing a thrombin cleavage site.
This unbound protein has several unique properties that can be incorporated into your own customized assays for the study of DNA methylation.
This unbound protein has several unique properties that can be incorporated into your own customized assays for the study of DNA methylation.
- Competitive elution of the GST-MBD protein or GST-MBD-methylated DNA complexes with glutathione.
- Elution of intact complexes by thrombin digestion
- Immobilization of protein DNA complexes or GST-MBD protein to other solid surfaces such as microtiter plates coated with GST antibodies.
- Visualization of complexes using labeled GST antibodies or GST antibodies and labeled secondary antibodies.
- Surface cysteine groups on the GST protein can be chemically modified to allow the addition of other reporters, affinity tags, or functional groups.
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Application) | For study of CpG methylation in DNA |
| Product Type | CpG MethylQuest™ Protein (Recombinant GST-MBD fusion protein) |
| Quantity | 100 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?