Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ CTCFα, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710044MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-100-44MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-44MI Supplier MilliporeSigma™ Supplier No. 1710044
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CTCFα Clone: in Human samples, and it is validated for Chromatin Immunoprecipitation, Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Multiplexing, Western Blotting

CTCF, also known as CCCTC-binding factor, contains 11 zinc finger (ZF) binding domains that allow it to bind different DNA target sequences and proteins. This multifunctional protein can act as a transcriptional activator or repressor. As a transcriptional repressor, CTCF, binds to promoters of vertebrate c-myc gene. It also binds to the PLK and PIM1 promoters. CTCF may prevent the access of transcriptional activators to enhancers. CTCF also acts as a transcriptional activator of APP. It is involved in different aspects of gene regulation including promoter activation or repression, hormone-responsive gene silencing, methylation-dependent chromatin insulation, and genomic imprinting. Mutations of CTCF in this gene have been associated with invasive breast cancers, prostate cancers, and Wilms tumors, suggesting a role for CTCF may as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTCFα
Applications Multiplex, Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoassay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-CTCF, (mouse monoclonal IgG1κ. One vial containing 50μg of protein G purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide, before the addition of glycerol to 30%. Normal Mouse IgG. Two vials containing 25μg purified IgG in 25μL storage buffer containing 0.1% sodium azide. ChIP Primers, Human H19 ICR. One vial containing 75μL of 5μM of each primer specific for human H19 internal control region. FOR: CCC ATC TTG CT GACC TCAC; REV: AGA CCT GGG ACG TTT CTG TG
Gene Accession No. P49711
Gene Symbols CTCF
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to human CTCF.
Purification Method Protein G purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_006556.1
Test Specificity Recognizes CTCF, Mr ∽130kDa.
Includes The ChIPAb+ CTCF set includes the CTCF antibody, a negative control antibody (normal mouse IgG) and qPCR primers which amplify a 165bp region of human H19 internal control region.
Target Species Human
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.