Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Dimethyl-Histone H3 (Lys27)α, Rabbit Monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710108MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-101-08MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-08MI Supplier MilliporeSigma™ Supplier No. 1710108
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

Specifically detects Dimethyl-Histone H3 (Lys27)α Clone: in Vertebrate samples, and it is validated for Chromatin Immunoprecipitation, Western Blotting

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome. Histones are modified post-translationally by the actions of enzymes in both the nucleus and cytoplasm. These modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications can alter local chromatin architecture, or recruit trans-acting factors that recognize specific histone modifications (the histone code hypothesis). The modifications occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle. Histone H3 is methylated at Lys27 by EZH2, and and overexpression of EZH2 has been associated with both breast and prostate cancers. Methylation of H3K27 is involved in X chroosome inactivation, imprinting, circadian rhythms, and stem cell maintenance. H3K27me2 is a marker of classical heterochromatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dimethyl-Histone H3 (Lys27)α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-Dimethyl-Histone H3 (Lys27) (rabbit monoclonal). One vial containing 50μL of cultured supernantant in 0.05% sodium azide. Negative Control Supernatant. One vial containing 100μL of rabbit cultured supernatant in 0.05% sodium azide. ChIP Primers, human β-globin. One vial containing 75μL of 5μM of each primer specific for the human β-globin promoter. FOR: AGG ACA GGT ACG GCT GTC ATC; REV: TTT ATG CCC AGC CCT GGC TC
Gene Accession No. Q71DI3
Gene Symbols H3, H3.2, H3/M, H3/m, H3/o, H3F2, H3FM, MGC9629, OTTHUMP00000014041
Host Species Rabbit
Immunogen Peptide containing the sequence (ARme2KSA) in which me2 corresponds to dimethyl lysine at residue 27 of human histone H3.
Purification Method Unpurified supernatant
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003484.1
Test Specificity This antibody recognizes Histone H3 dimethylated on Lys27.
Includes This ChIPAb+ Dimethyl-Histone H3 (Lys27) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Vertebrates
Content And Storage −20°C for one year from date of shipment
Form Culture Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.