Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Dimethyl-Histone H3 (Lys36), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710136MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-101-36MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-36MI Supplier MilliporeSigma™ Supplier No. 1710136
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects Dimethyl-Histone H3 (Lys36) Clone: in Human, Vertebrate samples, and it is validated for ChIP, Electromobility Shift Assay, Immunoprecipitation, Western Blotting

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histones are modified post-translationally by the actions of enzymes in both the nucleus and cytoplasm. These modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications can alter local chromatin architecture, or recruit trans-acting factors that recognize specific histone modifications (the histone code hypothesis). The modifications occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle. Methylation of histone H3 on Lys36 (H3K36me2/3) is tightly associated with actively transcribed genes, and this modification is found primarily within the coding region, suggesting H3K36 methylation is necessary for efficient RNA polymerase II elongation and processivity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dimethyl-Histone H3 (Lys36)
Applications ChIP Assay, Electromobility Shift Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Dimethyl Histone H3 (Lys36) (Rabbit Polyclonal). One vial containing 25μL rabbit polyclonal serum containing 0.05% sodium azide, before the addition of 30% glycerol. Normal Rabbit Serum. One vial containing 25μL antiserum containing 0.05% sodium azide. ChIP Primers, GAPDH coding D2. One vial containing 75μL of 5μM of each primer specific for human GAPDH coding region. (chr12:6647453+6647539, hg 19 build). FOR: 5’ GCC ATG TAG ACC CCT TGA AGA GC 3’; REV: 5’ ACT GGT TGA GCA CAG GGT ACT TTA T 3’
Gene Accession No. Q16695
Gene Symbols H3.4, H3/g , H3/t, H3FT, H3T, H3t, MGC126886, MGC126888, OTTHUMP00000037945
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a region near the N-terminus of human Histone H3 dimethylated at Lys36.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003484
Test Specificity Recognizes Dimethyl Histone H3 (Lys36). No detectable cross-reactivity unmethylated, mono, or dimethylated Lys36. No detectable cross reactivity to other histone modifications as measured by AbSurance Histone Antibody Specificity Arrays.
Includes This ChIPAb+ Validated Antibody and Primer Set conveniently includes the antibody, matched IgG negative control antibody and set control PCR primers that detect a known positive locus.
Target Species Human, Vertebrates
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.