Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Dimethyl-Histone H4 (Arg3) Symmetric, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710250MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-50MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-50MI Supplier MilliporeSigma™ Supplier No. 1710250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Dimethyl-Histone H4 (Arg3) Symmetric Clone: in Human, Mouse, Rat samples, and it is validated for ChIP, Dot Blot, Western Blotting

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Histones are modified post-translationally by the actions of enzymes in both the nucleus and cytoplasm. These modifications, which include acetylation, phosphorylation, methylation, and ubiquitin-ation, occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle. Arginine modifications of histones proteins are mediated by two classes of protein arginine methyltransferases (PRMTs), class I and II. Class I PRMTs place two methyl groups on the one nitrogen atom of the arginine guanidino group, resulting in an asymmetric modification. In contrast class II PRMTs place two methyl groups on two nitrogen atoms of the guanidino group producing a symmetric dimethylation. Although there are exceptions, asymmetric modifications are typically associated with transcription activation while symmetric modifications are most commonly associated with transcriptional repression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dimethyl-Histone H4 (Arg3) Symmetric
Applications ChIP Assay, Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Dimethyl-Histone H4 (Arg3) Symmetric (rabbit polyclonal). One vial containing 17.5μg of purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide before the addition of glycerol to 30%. Concentration: 0.7μg/μL. Normal Rabbit IgG. One vial containing 125μg of Rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide. ChIP Primers, human β-globin. One vial containing 75μL of 5μM of each primer specific for the human β-globin promoter. FOR: AGG ACA GGT ACG GCT GTC ATC; REV: TTT ATG CCC AGC CCT GGC TC
Gene Accession No. P62805
Gene Symbols HIST1H4A, H4/J, HIST1H4F, HIST1H4L, H4FN, H4FA, H4FH, HIST1H4H, H4F2, H4FM, H4/A, H4FK, H4FG, HIST2H4, H4FB, H4/K, HIST1H4I, H4/N, HIST1H4B, H4FE, H4/M, H4/a, HIST1H4E, HIST4H4, HIST2H4A, H4FC, H4FI, H4/H, HIST1H4C, H4/G, HIST1H4K, H4FJ, H4FD, H4/I, H4/B, H4/D, H4/C, HIST1H4J, HIST1H4D, H4/E
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Histone H4 when symmetrically dimethylated at Arg3.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001029249
Test Specificity This antibody recognizes Histone H4 symmetrically dimethylated at Arg3.
Includes This ChIPAb+ Dimethyl-Histone H4 (Arg3) Symmetric -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse, Rat
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.