Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Dkk-1, Rat, Recombinant Protein

Catalog No. 37120810UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-120-810UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-120-810UG Supplier MilliporeSigma™ Supplier No. 37120810UG
Add to Cart
Add to Cart
Recombinant, rat Dickkopf related protein-1 (Dkk-1), consisting of amino acids 23-270, fused at the C-terminus to a His·Tag™ sequence, and expressed in NS0-derived mouse myeloma cells. The balance between Wnt signaling and Dkk-1 inhibition is essential for bone formation and homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.2μM filtered PBS, BSA.
Preparation Method Reconstitute to 100μg/mL with sterile PBS containing ≥1% human or bovine serum albumin.
Purification Method SDS-PAGE
Quantity 10 μg
Immunogen Dkk-1
Storage Requirements Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C.
Regulatory Status RUO
Species Rat
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.