Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Dopamine D4 Receptor Control Peptide

Catalog No. AG222MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG222MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG222MI Supplier MilliporeSigma™ Supplier No. AG222
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Gene ID (Entrez) NM_000797.2
Concentration 0.2 mg/mL
Content And Storage -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Format Liquid
Formulation Liquid in PBS, pH 7.4 containing 0.1% sodium azide.
Gene Symbol DRD4, D4DR, D
Product Type Dopamine D4 Receptor, control peptide for AB1789/AB1789P
Quantity 100 μg
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.