Learn More
MilliporeSigma™ Dopaminergic Differentiation Growth Factor Sampler
Catalog No. SCR128 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SCR128 1 Kit
1 options
Catalog No. SCR128 Supplier MilliporeSigma™ Supplier No. SCR128
Add to Cart
Add to Cart
contains five validated growth factors used to induce differentiation of human pluripotent Embryonic stem (ES) and induced pluripotent Stem (iPS) cells to dopaminergic neurons.
TRUSTED_SUSTAINABILITY
Specifications
Purity or Quality Grade Greater than 98% by SDS-PAGE and HPLC analysis
Quantity 1 Kit