Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ E2F-3α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710062MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-100-62MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-62MI Supplier MilliporeSigma™ Supplier No. 1710062

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects E2F-3α Clone: in Human, Mouse, Rat samples, and it is validated for ChIP, Western Blotting

E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F 3 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F-3α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-E2F-3 (mouse monoclonal), . One vial containing 25μg of protein A purified mouse IgG2a in ∽180μL of storage buffer (10mM PBS, pH 7.4, 0.2% BSA, 0.09% sodium azide before addition of 30% glycerol). Normal Mouse IgG. One vial containing 25μg purified IgG in 25μL storage buffer containing 0.1% sodium azide. ChIP Primers, human CDC2 coding region, . One vial containing 75μL of 5μM each primer specific for human CDC2 coding region. FOR: CGC CCT TTC CTC TTT CTT TC; REV: ATC GGG TAG CCC GTA GAC TT
Gene Accession No. O00716
Gene Symbols E2F3, KIAA0075, E2F-3, MGC104598, DKFZp686C18211
Host Species Mouse
Immunogen Recombinant full-length human E2F-3 protein
Purification Method Protein A purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001940.1
Test Specificity E2F-3
Includes This ChIPAb+ E2F-3 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse, Rat
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.