Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ EED, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710034MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-100-34MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-34MI Supplier MilliporeSigma™ Supplier No. 1710034
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects EED Clone: in Bovine, Canine, Chicken, Human, Mouse, Opossum, Rat samples, and it is validated for ChIP, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen EED
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-EED (Rabbit polyclonal IgG). One vial containing 50μg of affinity purified antibody in 50μL of 0.1 M Tris-Glycine (pH 7.4) 150mM NaCl, containing 0.05% azide. Normal rabbit IgG. 1 vial containing 125μg of purified rabbit IgG in 125μL of storage buffer containing 0.1% sodium azide. ChIP Primers, HoxA2 upstream. One vial containing 75μL of 5μM of each primer specific for the promoter region of human HoxA2. FOR: AGG AAA GAT TTT GGT TGG GAA G; REV: AAA AAG AGG GAA AGG GAC AGA C
Gene Accession No. O75530
Gene Symbols EED, WAIT-1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a.a. 429-441 of human EED, conjugated to KLH.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003788
Test Specificity Recognizes EED, Mr 62 to 70kDa.
Includes This ChIPAb+ EED -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Bovine, Canine, Chicken, Human, Mouse, Opossum, Rat
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.