Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Elix™ 20 35 70 and 100 and RiOs™ 30 50 100 150 200 Pretreatment and Purification Packs

Catalog No. PRPK0L0S1US Shop All MilliporeSigma Products
Change view
Click to view available options
PrePak L1 Pre-System Pretreatment Pack for US, Canada and Mexico only
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRPK0L0S1US PrePak L1 Pre-System Pretreatment Pack for US, Canada and Mexico only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRPK0L0S1US Supplier MilliporeSigma™ Supplier No. PRPK0L0S1US

For Elix 20/35/70/100 and RiOs™ 35/50/100/150/200 water purification systems

Activated carbon and depth filtration; US, Canada and Mexico only
TRUSTED_SUSTAINABILITY

Specifications

Purification Method Activated Carbon, Depth Filtration
For Use With (Equipment) RiOs™ and Elix™ Water Purification System
Model PrePak L1
Type Pretreatment Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.