Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Elix™ 20/35/70/100 Water Purification Systems

Catalog No. ZLXS60035 Shop All MilliporeSigma Products
Change view
Click to view available options
110V 50/60Hz; 10A; Warning, contains mercury; Elix 35; Floerate: 35L/hr
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ZLXS60035 110V 50/60Hz; 10A; Warning, contains mercury; Elix 35; Floerate: 35L/hr
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. ZLXS60035 Supplier MilliporeSigma™ Supplier No. ZLXS60035

missing translation for 'validMainframeMsg'

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Requires

  • 8mm tubing for product and Elix module reject water
  • 1/2 in. tubing for RO reject water
  • >1.3 gal./min. (5L/min.) drain

Use Reverse Osmosis and electrodeionization Elix technology to meet pure water requirements for a lab or entire building

Produce up to thousands of L/day of Type II analytical-grade water from potable tap-water feed, <200μS/cm at 77°F (25°C), and provide total control of all operating parameters and monitoring functions both within the Elix system and for the external pure water distribution loop.

Key Features

  • Four-step purification systems support pure water production, storage, distribution and quality monitoring
  • Total control ensures constant flowrates and consistent water quality
  • Relevant parameters are checked after each purification step
  • Water quality at system outlet and in distribution loop is measured with flow-through resistivity cell designed to meet USP (645) requirements
  • Accuracy is assured with low 0.15cm-1 cell constant

Progard™ TL Pretreatment Pack Features

  • Pack removes particles (>0.5μm filter), free chlorine, and colloids (activated carbon filter) from tap water to protect system
  • Antiscaling agent protects reverse osmosis membrane in hard water areas
  • Bactericidal carbon filter prevents unwanted bacteria growth
  • Pretreatment packs are tailored to feedwater source and are available separately with or without chlorine tablets to autoclean RO membrane of bacteria and fouling contaminants
  • Three-way solenoid valve on RO membrane outlet closes product-outlet connection to system and directs any solution to drain during cleaning
  • Built-in TAG guarantees correct type of pack is used and warns user prior to exhaustion
Advanced Reverse Osmosis (RO) Membrane Features
  • Membrane removes 95-99% of inorganic ions and 99% of all dissolved organics (200 Daltons), microorganisms and particles
  • Reject water is recycled back to RO membrane feedwater stream
  • Water recovery can be adjusted from 35-75% to optimize water consumption, depending on feedwater quality and pretreatment sequence used
  • Constant flowrate at operating temperatures of 30° to 86°F (7° to 30°C) is maintained
  • RO module automatically sanitized when used with Progard TL Autoclean Pretreatment Pack
Elix Module Features
  • Module removes remaining ions by electrodeionization
  • Electrical current across module pulls ions captured in resin beds through cation- or anion-permeable membranes toward appropriate electrode
  • Ion-exchange resins are continuously regenerated by electric field applied within module—eliminating need to interrupt water production for hazardous chemical regeneration or costly resin replacement
  • Resins always remain at the highest quality and do not suffer degradation from exposure to harsh regeneration chemicals or removal
  • Activated carbon beads next to cathode lower pH at electrode surface to avoid scaling
  • Ions in concentrating channels are flushed from system into waste stream
UV Lamp Features
  • Wavelength: 254nm
  • Sanitizes water as last step
  • Log reduction value (LRV) of 4 in bacteria count (10,000CFU/mL is reduced to 1CFU/mL) seen irrespective of nominal flowrate—producing optimum water quality for bacteria-sensitive applications
System Operation Features
  • Operating modes and performance parameters displayed in one of six available languages on 4-line (20 character/line) back-lit display
  • Flashing LED and display message indicate when maintenance is required
  • Optional TOC Upgrade Kit (ZLX0 T0C KT) enables units to accurately measure and display total organic carbon (1 to 999ppb) of final product water or water in distribution loop
  • Systems permit direct control of distribution loop pump, inline loop UV lamp, Automatic Sanitization Module (ASM) on storage reservoir, water detector, automatic loop drain valve, and optional booster pump when tap feedwater pressure is <29psi (2 bar) (all available separately)
  • Systems also check feed pressure, feedwater conductivity, temperature, RO pressure, RO permeate conductivity, and RO membrane efficiency (% ion rejection)
  • Water quality data is recorded in real time for full traceability Feedwater inlet: 1/2 NPT

Applications:

  • Produce water suitable for such applications as microbiological media and buffer preparation, hydroponics, and chemical and biochemical reagent manufacturing
  • Can produce feedwater suitable for Milli-Q™ and Super-Q™ Ultrapure water polishing systems, and such laboratory equipment as washing machines, clinical analyzers, stability chambers, humidifiers, autoclaves, weathering test equipment and hydrogen gas generators

Compliance

Produce Type II water defined by CAP, ISO 3696/BS 3997 and CLSI; Meet purified water requirements of European and U.S. Pharmacopoeia and ASTM™

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CAP, ISO 3696/BS 3997, NCCLS, ASTM
Conductivity <0.2μS/cm
Daily Output 700 L
Feedwater Source Potable Tap Water
Flow Rate 35
Product Water Type Type 2 Analytical Grade
Resistivity >5
Total Organic Carbon <30
Type Pretreatment and Polishing Water Purification System
Feedwater Pressure 2 to 6 bar
Model Elix 35
Modules Elix™
pH 4 to 10
Power Supply 10 A
Temperature Range (English) 44.6°F to 86°F
Temperature Range (Metric) 7°C to 30°C
Dimensions (L x W x H) 28.87 x 26.1 x 28.9 in. (73.3 x 66.2 x 73.4 cm)
Length (English) 28.87 in.
Length (Metric) 73.3 cm
Width (English) 26.1 in.
Width (Metric) 66.2 cm
Height (English) 28.9 in.
Height (Metric) 73.4 cm
Depth (English) 17.4 in.
Depth (Metric) 44.1 cm
Weight (English) 106 lb.
Weight (Metric) 48 kg
Electrical Requirements 120 V 50/60 Hz
Voltage 120 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.