Learn More
MilliporeSigma™ Elix™ Essential and RiOs™ Essential Water Purification Systems: Automatic Sanitization Module
Catalog No. TANKASMES Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Elix/RiOs Essential Water Purification System
Catalog No. For Use With (Equipment)
TANKASMES Elix/RiOs Essential Water Purification System
1 options
Catalog No. TANKASMES Supplier MilliporeSigma™ Supplier No. TANKASMES
Add to Cart
Add to Cart
Accessories for Elix Essential 3, 5, 10, 15 and RiOs Essential 5, 8, 16, 24 Water Purification Systems
For use with EMD Millipore Elix Essential and RiOs Essential Water Purification Systems
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Elix/RiOs Essential Water Purification System
Type Sanitation Module
For Use With (Application) Reduces bacteria levels in 30/60/100L storage reservoir
Electrical Requirements 230 V, 50 Hz, Single Phase