Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EmbryoMax™ 2-Mercaptoethanol (100X)

Catalog No. ES007E Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES007E 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES007E Supplier MilliporeSigma™ Supplier No. ES007E
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

may be used for routine mouse embryonic stem cell culture applications.

The EmbryoMax 2-Mercaptoethanol (100X) is available in a 20mL format and may be used for routine mouse embryonic stem cell culture applications.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2-8°C up to 18 months
For Use With (Application) Embryonic Stem cell culture
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.