Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EmbryoMax™ ES Cell Qualified Fetal Bovine Serum

Catalog No. ES009B Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES009B 500 mL
ES009C 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES009B Supplier MilliporeSigma™ Supplier No. ES009B
Add to Cart
Add to Cart

growth supplement for cell culture media

The EmbryoMax ES Cell Qualified, US origin Fetal Bovine Serum (FBS) is a growth supplement for cell culture media. Prescreened for stem cell culture
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C up to 2 years
Quantity 500 mL
For Use With (Application) Stem cell culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.