Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EmbryoMax™ DMEM (1X), w/4.5g/L Glucose, 2.25g/L Sodium Bicarbonate

Catalog No. SLM220B Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SLM220B 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SLM220B Supplier MilliporeSigma™ Supplier No. SLM220B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for routine mouse embryonic stem cell culture applications.

  • Format: Liquid
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type DMEM Medium
With Additives Glucose, Sodium Bicarbonate
Without Additives No Glutamine, No Sodium Pyruvate
For Use With (Application) Stem cell culture
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.