Learn More
MilliporeSigma™ Endo-β-Galactosidase, Bacteroides fragilis, Recombinant Protein, E. coli
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | Sterile-filtered in 20mM Tris-HCl, pH 7.5. |
| Quantity | 50 μL |
| Source | E. coli |
| Immunogen | Endo-β-Galactosidase |
| Storage Requirements | There is no significant loss of activity if left at ambient temperature for several days. The enzyme is active for at least 5 days under the Recommended Reaction Conditions above. |
| Regulatory Status | RUO |
| Species | B. fragilis |
| Recombinant | Recombinant |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.