Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Endoglycosidase H, Streptomyces plicatus, Recombinant Protein, E. coli

Catalog No. 324717200MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 mIU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-471-7200MI 200 mIU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-471-7200MI Supplier MilliporeSigma™ Supplier No. 324717200MIU
Add to Cart
Add to Cart
Recombinant, streptomyces plicatus endoglycosidase H expressed in E. coli. Catalyzes the hydrolysis of asparagine-linked oligomannose and hybrid, but not complex oligosaccharides from glycoprotein. Endo H cleaves between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 50mM NaCl, 20mM Tris-HCl, 1mM EDTA, pH 7.5.
Quantity 200 mIU
Source E. coli
Immunogen Endoglycosidase H
Regulatory Status RUO
Species S. plicatus
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.