Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Epidermal Growth Factor (EGF), Human Recombinant Protein

Catalog No. 01107MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-107MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-107MI Supplier MilliporeSigma™ Supplier No. 01107
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

For Use With (Application) Cell Culture
Name Epidermal Growth Factor (EGF)
Quantity 100 μg
Source Recombinant DNA expressed in yeast (S. cerevisiae)
Cross Reactivity Human
Recombinant Recombinant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.