Learn More
MilliporeSigma™ ES2N Basal Medium
Catalog No. SCM083 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
245 mL
Catalog No. Quantity
SCM083 245 mL
1 options
Catalog No. SCM083 Supplier MilliporeSigma™ Supplier No. SCM083
Add to Cart
Add to Cart
a defined serum-free neural basal medium.
ES2N Basal Medium is a defined serum-free neural basal medium. This base medium offers flexibility to customers adding their own neural supplements or growth factors. Must be supplemented prior to use.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Stem cell culture
Quantity 245 mL