Learn More
MilliporeSigma™ ES2N Complete Medium Kit
Catalog No. SCM082 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
Catalog No. Quantity
SCM082 250 mL
1 options
Catalog No. SCM082 Supplier MilliporeSigma™ Supplier No. SCM082
Add to Cart
Add to Cart
a defined serum-free formulation that has been optimized for the differentiation of mouse embryonic stem (ES) and induced pluripotent stem (iPS) cells into neurons.
ES2N Complete medium is a defined serum-free formulation that has been optimized for the differentiation of mouse embryonic stem (ES) and induced pluripotent stem (iPS) cells into neurons. When used in conjucture with ESGRO Complete Plus medium mouse ES and iPS cells readily differentiate into neurons in a monolayer assay within 9-12 days on gelatin coated culture dishes. When supplemented with bFGF and EGF, this medium enables the isolation and expansion of neural stem cells (NSCs).
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Stem cell culture
Quantity 250 mL