Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ESGRO Complete™ Serum-Free Cell Culture Freezing Medium

Catalog No. SF005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SF005 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SF005 Supplier MilliporeSigma™ Supplier No. SF005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

available in a 50mL format and has been optimized and validated for Stem cell culture.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Stem Cell Culture
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.