Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Estrogen Receptor αα, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17603MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-603-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-603-MI Supplier MilliporeSigma™ Supplier No. 17603
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects Estrogen Receptor αα Clone: in Human, Mouse samples, and it is validated for ChIP, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Estrogen Receptor αα
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-ERα (mouse ascites). 1 vial containing 100mL ascites. The ERα antibody is made against aa120-170 of bovine estrogen a. It can recognize human and mouse ERα. ChIP primers TFF1 (pS2). 1 vial containing 75mL of 5μM of each control primer specific for human TFF1 (pS2) promoter. FOR: CCG GCC ATC TCT CAC TAT GAA; REV: CCT TCC CGC CAG GGT AAA TAC
Gene Accession No. P03372
Gene Symbols ESR1, Era, ER-alpha, ESR, ER, NR3A1, ESRA, DKFZp686N23123
Host Species Mouse
Immunogen The ERα antibody is made against aa 120-170 of bovine estrogen α.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_000125.2
Test Specificity Estrogen Receptor α
Includes This ChIPAb+ Estrogen Receptor α -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage −20°C for one year from date of shipment
Form Ascites
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.