Learn More
MilliporeSigma™ External Relay Kit
Catalog No. KITRELESS Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Elix/RiOs Essential Water Purification System
Catalog No. For Use With (Equipment)
KITRELESS Elix/RiOs Essential Water Purification System
1 options
Catalog No. KITRELESS Supplier MilliporeSigma™ Supplier No. KITRELESS
Add to Cart
Add to Cart
For Elix™ Essential and RiOs™ Essential Water Systems
  • Allows users to send system alerts and alarms to a remote monitoring device or display
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Elix/RiOs Essential Water Purification System
Type Kit
For Use With (Application) Allows users to send system alerts and alarms to a remote monitoring device or display