Learn More
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Molecular Weight (g/mol) | 17.5kDa |
| Name | Fibroblast Growth Factor (FGF) |
| Quantity | 25 μg |
| Source | Recombinant DNA expressed in yeast (S. cerevisiae) |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Purity or Quality Grade | Greater than 96% as determined by HPLC and SDS-PAGE prior to the addition of BSA and CHAPS |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.