Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Fibrinogen, Plasminogen-Depleted, Human Plasma, Protein

Catalog No. 341578500MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-157-8500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-157-8500MG Supplier MilliporeSigma™ Supplier No. 341578500MG
Add to Cart
Add to Cart
Native fibrinogen from human plasma, depleted of plasminogen using a Lys-agarose column. An acute-phase plasma protein found at about 300mg/dL. Contains ≥95% clottable proteins.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 20mM sodium citrate-HCl, pH 7.4.
Preparation Method H2O
Purification Method Homogeneous by SDS-PAGE
Quantity 500 mg
Immunogen Fibrinogen
Storage Requirements Following reconstitution, aliquot and freeze (−70°C) for long term storage or store at room temperature (20°C) for short-term storage. Stock solutions are stable for up to 4hr. at 20°C or for up to 6 months at −70°C. Do not store solutions on ice; precipitation will occur. Stock solutions stored at −70°C should be thawed in a water bath maintained at 37°C.
Regulatory Status RUO
Without Additives Plasminogen-Depleted
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade Clottable proteins: ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.