Learn More
MilliporeSigma™ Adapter, 1/4 in. NPTF to F Luer-Lok
Catalog No. XX3002568 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Catalog No. For Use With (Equipment)
XX3002568 Water Purification System
1 options
Catalog No. XX3002568 Supplier MilliporeSigma™ Supplier No. XX3002568
Add to Cart
Add to Cart
Accessories for filter holders for sample preparation.
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
TRUSTED_SUSTAINABILITY
Specifications
Connection Type NPTF to F
For Use With (Equipment) Water Purification System
Type Laboratory Filter Holders or Cyclones
Material Nickel/Chrome plated
Diameter (English) Inside 0.25 in.
Diameter (Metric) Inside 0.63 cm