Description
Includes
Glass filter holder (47mm) with stainless steel screen, 1L Vacuum filtering flask, blunt end filter forceps, latex tubing (1/4 in. x 23 in.), 25mm Solvent Filtering Dispenser, 400 PetriSlides, Chemical Duty Pump (115V), Chemical Duty Pump (220V).
Specifications
Specifications
| Certifications/Compliance | ASTM-D2276, ARP-598 |
| Material | Stainless Steel |
| Includes | Vacuum/pressure pump, 47 mm stainless steel filter holder, 1 L flask, forceps, 0.25 in. I.D. tubing, 25 mm solvent dispenser, PetriSlides, filters, chemical duty pump |
| Type | Fluids Contamination Analysis Kit |