Learn More
MilliporeSigma™ Fluorescein-5-isothiocyanate (FITC) Dye
Catalog No. M1245460250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
Catalog No. Quantity
M1245460250 250 mg
1 options
This item is not returnable. View return policy
Catalog No. M1245460250 Supplier MilliporeSigma™ Supplier No. 1.24546.0250
This item is not returnable. View return policy
Fluorescein-5-isothiocyanate (FITC) for biochemistry and for fluorescence microscopy. CAS 3326-32-7, molar mass 389.38g/mol.
In immuno histology/ cytology, Fluorescein-5-isothiocyanate (FITC) for biochemistry and for fluorescence microscopy, serve as marker substance for the visualization and for the detection of tissue-bound bio molecules like e.g. antigens, lectins, various proteins, peptides, nucleic acids, oligo- and polysaccharides in samples of human origin. FITC-marked antibodies bind specifically to the appropriate target structures in the tissue. An optimum FITC-protein (antibody) ratio is important for a good result.
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Quantity 250 mg
Product Type Fluorescent Dye