Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ FOXA1, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710267MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-67MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-67MI Supplier MilliporeSigma™ Supplier No. 1710267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects FOXA1 Clone: in Human, Mouse samples, and it is validated for Chromatin Immunoprecipitation, Immunocytochemistry, Western Blotting

HNF-3A, Hepatocyte nuclear factor 3-alpha (also called FOXA1) is member of the Forkhead family of winged-helix transcription factors. This protein family has around 50 known transcription factors and play an important role in the regulation of cell differentiation, organogenesis, and gene expression. High levels of expression of HNF-3A has been observed in tumors arising from both the prostate and breast. Here it is believed this protein interacts with the androgen receptor in prostate tumors and the estrogen receptor α in breast tumors. In the case of ERα, HNF-3A interacts with cis-regulatory regions in heterochromatin and enhances the interaction of ERα with chromatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXA1
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-FOXA1 (Rabbit Polyclonal). One vial containing 50μL of purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide before the addition of glycerol to 30%. Concentration: 0.35mg/mL. Normal Rabbit IgG. One vial containing 125μg Rabbit IgG in 125μL storage buffer containing 0.05% sodium azide. ChIP Primers, Mouse Hnf4α enhancer. One vial containing 75μL of 5μM of each primer specific for Mouse Hnf4α enhancer. FOR: TTC CAG CTG CCT TTA TCT CCC TGT; REV: TCT CCA CAC ATG TCC AGC AGC CT
Gene Accession No. P55317
Gene Symbols FOXA1, HNF-3-alpha, HNF-3A, HNF3A, TCF3A, MGC33105
Host Species Rabbit
Immunogen Recombinant protein corresponding to the entire protein of human FOXA1
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_004487
Test Specificity This antibody recognizes HNF-3A/FOXA1.
Includes This ChIPAb+ FOXA1 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.