Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ FOXA2α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710258MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-58MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-58MI Supplier MilliporeSigma™ Supplier No. 1710258
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects FOXA2α Clone: in Human, Mouse samples, and it is validated for Chromatin Immunoprecipitation, Immunocytochemistry, Western Blotting

Also known as hepatocyle nuclear factor 3-beta (HNF-3B), Forkhead box protein A2 (FOXA2) belongs to the forkhead class of DNA-binding proteins. This family of transcription factors influences gene expression in endodermally-derived tissues such as lung, liver, stomach, pancreas, and intestine. Foxa2 has also been detected in ectodermally-derived tissues such as kidney, uterus and vagina, and seminal and coagulating glands during development. Several studies show that FOXA proteins help facilitiate the binding of several nuclear receptors to their respective targets in a context-dependent manner, and are thereby implicated in various functions including cellular maintenance, differentiation, metabolism, and organogenesis. Aberrant function of FOXA proteins are attributed to diseases states such as diabetes and Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXA2α
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-FOXA2 (mouse monoclonal),. One vial containing 100μL of purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4) and 150mM NaCl with 0.05% sodium azide before the addition of glycerol to 30%. Concentration: 0.175mg/mL. Normal Mouse IgG. One vial containing 125μg of purified mouse IgG in 125μL of storage buffer containing 0.1% sodium azide. ChIP Primers, Mouse Hnf4α enhancer. One vial containing 75μL of 5μM of each primer specific for Mouse Hnf4α enhancer. FOR: TTC CAG CTG CCT TTA TCT CCC TGT; REV: TCT CCA CAC ATG TCC AGC AGC CT
Gene Accession No. Q9Y261
Gene Symbols FOXA2, HNF3B, TCF3B
Host Species Mouse
Immunogen Recombinant protein corresponding to the human FOXA2.
Purification Method Protein G
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_068556
Includes This ChIPAb+ FOXA2 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.