Learn More
Specifications
Specifications
| Gene ID (Entrez) | NM_000818.1 |
| Content And Storage | Long term storage at -20°C in a sealed container with desiccant for up to 12 months after date of receipt. |
| Format | Lyophilized |
| Formulation | White lyophilized powder. Soluble in water. |
| Gene Symbol | GAD2, MGC161607, GAD65, MGC161605, GAD-65, EC 4.1.1.15 |
| Product Type | GAD, control peptide for AB1511 |
| Quantity | 50 μg |
| Regulatory Status | RUO |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.