Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ β-Galactosidase, E. coli, Protein

Catalog No. 3457885MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-578-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-578-85MG Supplier MilliporeSigma™ Supplier No. 3457885MG
Add to Cart
Add to Cart
Native E. coli β-galactosidase. Inducible enzyme not found in eukaryotic cells that is used as a reporter gene. Used to assess efficiency of transfection. Also used in ELISA and immunocytochemical techniques. Histochemical substrates include X-Gal.
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method H2O (2%)
Quantity 5 mg
Source E. coli
Immunogen β-Galactosidase
Storage Requirements Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C.
Regulatory Status RUO
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.