Learn More
MilliporeSigma™ Glucose Oxidase, Aspergillus niger, Recombinant Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Preparation Method | 0.1M Phosphate buffer, pH 7.0 (5mg/mL) |
| Quantity | 10 kU |
| Source | Aspergillus niger |
| Immunogen | Glucose Oxidase |
| Storage Requirements | Following reconstitution, aliquot and refrigerate (4°C). Stock solutions are stable for up to 3 months at 4°C. |
| Regulatory Status | RUO |
| Recombinant | Recombinant |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.