Learn More
MilliporeSigma™ Glutamate Receptor δ 1/2 Control Peptide for AB1514
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Gene ID (Entrez) | NM_017551.1 |
| Content And Storage | -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
| Format | Liquid |
| Formulation | Liquid |
| Gene Symbol | GRID1, KIAA1220, OTTHUMP00000059648 |
| Product Type | Glutamate Receptor δ 1/2 Control Peptide for AB1514 |
| Quantity | 100 μg |
| Regulatory Status | RUO |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.