Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ GPT Selection Reagent Kit

Catalog No. TR1001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TR1001 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TR1001 Supplier MilliporeSigma™ Supplier No. TR1001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

an antibiotic useful in research for the selection of animal cells that express the E. coli.

Mycophenolic Acid is an antibiotic useful in research for the selection of animal cells that express the
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 500X Mycophenolic Acid containing ETOH (12.5mg/mL) 2 x 1mL
1 bottle (10mL) of 100X Aminopterin solution which consists of: xanthine 25 mg, hypoxanthine: 1.5 mg, aminopterin: 0.2mg, 0.5M NaOH: 1mL vial. Store at -20°C
For Use With (Application) In research for selection of animal cells that express E. coli
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.