Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Granzyme A, Human, Recombinant Protein, E. coli

Catalog No. 36804410UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-804-410UG 10μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-804-410UG Supplier MilliporeSigma™ Supplier No. 36804410UG
Add to Cart
Add to Cart
Recombinant, human granzyme A expressed in E. coli. Granzyme A is a trypsin-like serine protease secreted by cytotoxic T lymphocytes (CTL) and natural killer (NK) cells. A proapoptotic enzyme that induces cell death through caspase-independent mechanism, causing single-stranded DNA nicking and other apoptotic features including nuclear fragmentation, chromatin condensation, loss of mitochondrial potential, and membrane blebbing. Granzyme A is also known to bind to and cleave nucleolin and activate the thrombin receptor on neuronal cells and astrocytes. It also cleaves and activates interleukin-1β. Note: The M.W. is that of a homodimer.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 520mM NaCl, 50mM MES, 10% glycerol, pH 6.0.
Quantity 10μg
Source E. coli
Immunogen Granzyme A
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.