Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ GSK-3β, Rabbit Skeletal Muscle, Recombinant Protein, E. coli

Catalog No. 3615265KU Shop All MilliporeSigma Products
Change view
Click to view available options
5kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
36-152-65KU 5kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-152-65KU Supplier MilliporeSigma™ Supplier No. 3615265KU
Only null left
Add to Cart
Add to Cart
Recombinant, rabbit glycogen synthase kinase 3β expressed in E. coli. Dual specificity kinase. One of several protein kinases that phosphorylates glycogen synthase. Other substrates include p90rsk, Tau, c-Jun, and CREB. Plays a key role in Wnt and insulin signaling.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 50mM NaCl, 30mM Tris-HCl, 5mM DTT, 1mM EDTA, 50% glycerol, 0.03% BRIJ™-35 Detergent, pH 7.5
Quantity 5kU
Source E. coli
Immunogen GSK-3β
Storage Requirements Following initial thaw aliquot and freeze (−20°C).
Regulatory Status RUO
Species Rabbit
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.