Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Harleco™ Fucillo Wright Giemsa Solution

Catalog No. 23750000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-750000 1 L
23-750001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-750000 Supplier MilliporeSigma™ Supplier No. 6457175
This item is not returnable. View return policy

Harleco™ Fucillo Wright Giemsa Solution for red blood cell morphology and white blood cell differentiation. For use with the MIDAS™ III Automated Stainer.

  • Polychromatic stain for peripheral blood and bone marrow smears
  • Used for red blood cell morphology and white blood cell differentiation
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 L
Boiling Point 64.5°C
Format Liquid
Type Wright Giemsa Solution
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.