Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Harleco™ Giemsa Stain Solution

Catalog No. 23426520 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-426520 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-426520 Supplier MilliporeSigma™ Supplier No. 620G75
This item is not returnable. View return policy

For use with the MIDAS™ III Automated Stainer

Stains G-bands of chromosomes in genetic studies, can be used with Wright Stain for blood and bone marrow staining

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 L
Format Original Azure Blend
Product Type Giemsa Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.